Please login to your online account to display your discounted pricing

Methyl 2,5-dimethyl-1H-pyrrole-3-carboxylate, 95%, Maybridge

Chemical Identifiers

Chemical Name or Material Methyl 2, 5-dimethyl-1H-pyrrole-3-carboxylate
CAS 69687-80-5
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C8H11NO2
Formula Weight 153.18
MDL Number MFCD00203859
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SP00004DA

  SDS
fisher scientific company
SP00004DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 153.18
Quantity 1g
Assay 95%
CAS Min % 95.0
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 95%
Chemical Name or Material Methyl 2, 5-dimethyl-1H-pyrrole-3-carboxylate
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
MDL Number MFCD00203859
Packaging Amber Glass Bottle
Molecular Formula C8H11NO2
CAS 69687-80-5