Please login to your online account to display your discounted pricing

Methyl 2,5-dimethyl-1H-pyrrole-3-carboxylate, 95%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  SP00004DA

 View more versions of this product

Catalog No. SP00004DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 153.18
Quantity 1g
Assay 95%
CAS Min % 95.0
Assay Percent Range 95%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Methyl 2, 5-dimethyl-1H-pyrrole-3-carboxylate
Molecular Formula C8H11NO2
MDL Number MFCD00203859
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Packaging Amber Glass Bottle
CAS 69687-80-5

Specifications

Chemical Name or Material Methyl 2, 5-dimethyl-1H-pyrrole-3-carboxylate
CAS 69687-80-5
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C8H11NO2
Formula Weight 153.18
MDL Number MFCD00203859
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.