Please login to your online account to display your discounted pricing

Methyl 2,5-dihydroxycinnamate, Tocris


EGFR-kinase inhibitor

Quantity 10mg

Manufacturer: r & d systems  RDS057710

 View more versions of this product

Catalog No. 057710

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 194.19
Chemical Name or Material Methyl 2, 5-dihydroxycinnamate
Assay Percent Range >98%
Molecular Formula C10H10O4
CAS 63177-57-1
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Methyl 2, 5-dihydroxycinnamate
Assay Percent Range >98%
Molecular Formula C10H10O4
Formula Weight 194.19
Accessories

Store at Room Temperature