Please login to your online account to display your discounted pricing

Methyl 2,5-dihydroxycinnamate, Tocris

EGFR-kinase inhibitor

$126.32 - $534.65

Chemical Identifiers

Chemical Name or Material Methyl 2, 5-dihydroxycinnamate
Assay Percent Range >98%
Molecular Formula C10H10O4
Formula Weight 194.19
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

057710

 
r & d systems
RDS057710
10mg Each for $126.32

057750

 
r & d systems
RDS057750
50mg Each for $534.65
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 194.19
Chemical Name or Material Methyl 2, 5-dihydroxycinnamate
Assay Percent Range >98%
Molecular Formula C10H10O4
CAS 63177-57-1
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature