missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-(5-chlorobenzo[b]thiophen-3-yl)acetate, 97%, Maybridge

Chemical Identifiers

CAS 95834-67-6
CAS Min % 97.0
Assay Percent Range 90%
Molecular Formula C11H9ClO2S
Formula Weight 240.71
Chemical Name or Material Methyl 2-(5-chlorobenzo[b]thiophen-3-yl)acetate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MWP00091DA SDS Fisher Scientific Company
MWP00091DA
Amber Glass Bottle 1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

95834-67-6
90%
240.71
97.0
C11H9ClO2S
Methyl 2-(5-chlorobenzo[b]thiophen-3-yl)acetate

Specifications

240.71
97.0
C11H9ClO2S
90%
1g
Amber Glass Bottle
95834-67-6
Methyl 2-(5-chlorobenzo[b]thiophen-3-yl)acetate

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.