missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 2,4-Dimethoxybenzoate 98.0+%, TCI America™
$101.21 - $101.21
Chemical Identifiers
CAS | 2150-41-6 |
---|---|
Molecular Formula | C10H12O4 |
Molecular Weight (g/mol) | 196.20 |
MDL Number | MFCD00070649 |
InChI Key | IHIJFZWLGPEYPJ-UHFFFAOYSA-N |
PubChem CID | 137451 |
IUPAC Name | methyl 2,4-dimethoxybenzoate |
SMILES | COC(=O)C1=CC=C(OC)C=C1OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
M22265G
|
TCI America
M22265G |
5 g |
Each for $101.21
|
|
|||||
Chemical Identifiers
2150-41-6 | |
196.20 | |
IHIJFZWLGPEYPJ-UHFFFAOYSA-N | |
methyl 2,4-dimethoxybenzoate |
C10H12O4 | |
MFCD00070649 | |
137451 | |
COC(=O)C1=CC=C(OC)C=C1OC |
Specifications
2150-41-6 | |
296°C | |
MFCD00070649 | |
2,4-dimethoxybenzoic acid methyl ester, benzoic acid, 2,4-dimethoxy-, methyl ester, acmc-1ccii, methyl2,4-dimethoxybenzoate, methyl 2,4-dimethoxybenzoate, 2,4-dimethoxy-benzoic acid methyl ester, benzoic acid,2,4-dimethoxy-,methyl ester, benzoic acid,2,4-dimethoxy-, methyl ester, 2,4-dihydroxybenzoic acid, dimethyl ether, methyl ester | |
COC(=O)C1=CC=C(OC)C=C1OC | |
196.20 | |
196.20 | |
Liquid |
Yellow | |
C10H12O4 | |
5 g | |
IHIJFZWLGPEYPJ-UHFFFAOYSA-N | |
methyl 2,4-dimethoxybenzoate | |
137451 | |
≥98.0% (GC) | |
Methyl 2,4-Dimethoxybenzoate |
Safety and Handling
TSCA : No