missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2,4-Dimethoxybenzoate 98.0+%, TCI America™

Manufacturer:  Tci America M222625G

 View more versions of this product

Catalog No. M222625G



Specifications

Specifications

Methyl 2,4-Dimethoxybenzoate
Yellow
2150-41-6
MFCD00070649
IHIJFZWLGPEYPJ-UHFFFAOYSA-N
methyl 2,4-dimethoxybenzoate
137451
Clear Liquid at 20°C
296°C
25g
C10H12O4
2,4-dihydroxybenzoic acid, dimethyl ether, methyl ester, 2,4-dimethoxy-benzoic acid methyl ester, 2,4-dimethoxybenzoic acid methyl ester, acmc-1ccii, benzoic acid, 2,4-dimethoxy-, methyl ester, benzoic acid,2,4-dimethoxy-, methyl ester, benzoic acid,2,4-dimethoxy-,methyl ester, methyl 2,4-dimethoxybenzoate, methyl2,4-dimethoxybenzoate
COC1=CC(=C(C=C1)C(=O)OC)OC
196.202
196.20
≥98.0% (GC)

Chemical Identifiers

2150-41-6
196.202
IHIJFZWLGPEYPJ-UHFFFAOYSA-N
137451
COC1=CC(=C(C=C1)C(=O)OC)OC
C10H12O4
MFCD00070649
2,4-dihydroxybenzoic acid, dimethyl ether, methyl ester, 2,4-dimethoxy-benzoic acid methyl ester, 2,4-dimethoxybenzoic acid methyl ester, acmc-1ccii, benzoic acid, 2,4-dimethoxy-, methyl ester, benzoic acid,2,4-dimethoxy-, methyl ester, benzoic acid,2,4-dimethoxy-,methyl ester, methyl 2,4-dimethoxybenzoate, methyl2,4-dimethoxybenzoate
methyl 2,4-dimethoxybenzoate
Safety and Handling

Safety and Handling

TSCA : No

Documents
Promotions

Promotions