Please login to your online account to display your discounted pricing

Methyl 2,4-dimethoxybenzoate, 98%, Acros Organics

Chemical Identifiers

Chemical Name or Material Methyl 2, 4-dimethoxybenzoate
Assay Percent Range 98%
Molecular Formula C10H12O4
Formula Weight 196.2
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC426810050

  SDS
thermo fisher scientific
AC426810050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC426811000

  SDS
thermo fisher scientific
AC426811000
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC426810250

  SDS
thermo fisher scientific
AC426810250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 196.2
Quantity 5g
Molecular Formula C10H12O4
Chemical Name or Material Methyl 2, 4-dimethoxybenzoate
CAS 2150-41-6
Packaging Glass bottle
Assay Percent Range 98%