Please login to your online account to display your discounted pricing

Methyl 2,4-dimethoxybenzoate, 98%, Acros Organics


Quantity 100g

Manufacturer: thermo fisher scientific  AC426811000

 View more versions of this product

Catalog No. AC426811000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 196.2
Quantity 100g
Molecular Formula C10H12O4
CAS 2150-41-6
Packaging Glass bottle
Chemical Name or Material Methyl 2, 4-dimethoxybenzoate
Assay Percent Range 98%

Specifications

Chemical Name or Material Methyl 2, 4-dimethoxybenzoate
Assay Percent Range 98%
Molecular Formula C10H12O4
Formula Weight 196.2