Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2,4-dimethoxy-6-methylbenzoate, 98%

$227.81 - $227.81

Chemical Identifiers

Chemical Name or Material Methyl 2,4-dimethoxy-6-methylbenzoate
CAS 6110-37-8
Molecular Formula C11H14O4
Formula Weight 210.23
MDL Number MFCD00082782
Beilstein 1372278
Synonym 2,4-Dimethoxy-6-methylbenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0989003

 
alfa aesar™
L0989003
1g Each for $227.81
Description & Specifications

Specifications

Formula Weight 210.23
Assay 98%
Molecular Formula C11H14O4
MDL Number MFCD00082782
CAS 6110-37-8
Beilstein 1372278
Synonym 2,4-Dimethoxy-6-methylbenzoic acid methyl ester
Quantity 1g
Chemical Name or Material Methyl 2,4-dimethoxy-6-methylbenzoate
Melting Point 48° to 50°C