Alfa Aesar™ Methyl 2,4-dimethoxy-6-methylbenzoate, 98%
Manufacturer:  Alfa Aesar™ L09890MD
 View more versions of this product
Catalog No. AAL09890MD
Specifications
Specifications
Methyl 2,4-dimethoxy-6-methylbenzoate
250mg
C11H14O4
1372278
QCEODMDPMRZZDI-UHFFFAOYSA-N
methyl 2,4-dimethoxy-6-methylbenzoate
601782
98%
48°C to 50°C
6110-37-8
MFCD00082782
acmc-20admr, o-toluic acid, 4,6-dimethoxy-, methyl ester, benzoic acid, 2,4-dimethoxy-6-methyl-, methyl ester, methyl 2,4-dimethoxy-6-methyl-benzoate, 4,6-dimethoxy-o-toluic acid methyl ester
CC1=CC(=CC(=C1C(=O)OC)OC)OC
210.229
210.23
Chemical Identifiers
6110-37-8
210.229
QCEODMDPMRZZDI-UHFFFAOYSA-N
601782
CC1=CC(=CC(=C1C(=O)OC)OC)OC
C11H14O4
MFCD00082782
acmc-20admr, o-toluic acid, 4,6-dimethoxy-, methyl ester, benzoic acid, 2,4-dimethoxy-6-methyl-, methyl ester, methyl 2,4-dimethoxy-6-methyl-benzoate, 4,6-dimethoxy-o-toluic acid methyl ester
methyl 2,4-dimethoxy-6-methylbenzoate
Documents