Please login to your online account to display your discounted pricing

Methyl 2,4-dihydroxybenzoate, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  126730250

 View more versions of this product

Catalog No. AC126730250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 168.15
Physical Form Powder
Assay ≥96.0%
Quantity 25g
Color White to beige
Melting Point 116° to 121°C
Linear Formula (HO)2C6H3CO2CH3
Assay Percent Range 97%
CAS 2150-47-2
Molecular Formula C8H8O4
Chemical Name or Material Methyl 2, 4-dihydroxybenzoate
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00002276
Packaging Glass bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Methyl 2, 4-dihydroxybenzoate
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C8H8O4
Linear Formula (HO)2C6H3CO2CH3
Formula Weight 168.15
MDL Number MFCD00002276
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents