Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2,4-difluorobenzoate, 98%

Chemical Identifiers

Chemical Name or Material Methyl 2,4-difluorobenzoate
CAS 106614-28-2
Molecular Formula C8H6F2O2
Formula Weight 172.13
MDL Number MFCD03093799
Synonym 2,4-Difluorobenzoic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3216114

  SDS
alfa aesar™
H3216114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 172.13
Quantity 25g
Chemical Name or Material Methyl 2,4-difluorobenzoate
Synonym 2,4-Difluorobenzoic acid methyl ester
CAS 106614-28-2
Assay 98%
Molecular Formula C8H6F2O2
MDL Number MFCD03093799