missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 2,4-Dichlorophenylacetate 99.0+%, TCI America™
$66.37 - $199.08
Chemical Identifiers
CAS | 55954-23-9 |
---|---|
Molecular Formula | C9H8Cl2O2 |
Molecular Weight (g/mol) | 219.061 |
MDL Number | MFCD00018381 |
InChI Key | SRCVNBFTEARRJY-UHFFFAOYSA-N |
Synonym | methyl 2,4-dichlorophenylacetate, methyl 2-2,4-dichlorophenyl acetate, 2,4-dichlorophenylacetic acid methyl ester, benzeneacetic acid, 2,4-dichloro-, methyl ester, methyl2,4-dichlorophenylacetate, dcaa methyl ester, acmc-209lq7, methyl 2,4-dichlorobenzeneacetate, 2,4-dichlorophenylaceticacidmethylester, 2,4-dichlorobenzeneacetic acid methyl ester |
PubChem CID | 2734106 |
IUPAC Name | methyl 2-(2,4-dichlorophenyl)acetate |
SMILES | COC(=O)CC1=C(C=C(C=C1)Cl)Cl |
Chemical Identifiers
55954-23-9 | |
219.061 | |
SRCVNBFTEARRJY-UHFFFAOYSA-N | |
2734106 | |
COC(=O)CC1=C(C=C(C=C1)Cl)Cl |
C9H8Cl2O2 | |
MFCD00018381 | |
methyl 2,4-dichlorophenylacetate, methyl 2-2,4-dichlorophenyl acetate, 2,4-dichlorophenylacetic acid methyl ester, benzeneacetic acid, 2,4-dichloro-, methyl ester, methyl2,4-dichlorophenylacetate, dcaa methyl ester, acmc-209lq7, methyl 2,4-dichlorobenzeneacetate, 2,4-dichlorophenylaceticacidmethylester, 2,4-dichlorobenzeneacetic acid methyl ester | |
methyl 2-(2,4-dichlorophenyl)acetate |
Specifications
55954-23-9 | |
153°C | |
MFCD00018381 | |
methyl 2,4-dichlorophenylacetate, methyl 2-2,4-dichlorophenyl acetate, 2,4-dichlorophenylacetic acid methyl ester, benzeneacetic acid, 2,4-dichloro-, methyl ester, methyl2,4-dichlorophenylacetate, dcaa methyl ester, acmc-209lq7, methyl 2,4-dichlorobenzeneacetate, 2,4-dichlorophenylaceticacidmethylester, 2,4-dichlorobenzeneacetic acid methyl ester | |
COC(=O)CC1=C(C=C(C=C1)Cl)Cl | |
219.061 | |
219.06 | |
Liquid |
Yellow | |
C9H8Cl2O2 | |
5 g | |
SRCVNBFTEARRJY-UHFFFAOYSA-N | |
methyl 2-(2,4-dichlorophenyl)acetate | |
2734106 | |
≥99.0% (GC) | |
Methyl 2,4-Dichlorophenylacetate |
Safety and Handling
TSCA : No