missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 2,4-Dichlorophenylacetate 99.0+%, TCI America™
Supplier: TCI America M252825G
Specifications
Methyl 2,4-Dichlorophenylacetate | |
Yellow | |
C9H8Cl2O2 | |
25 g | |
SRCVNBFTEARRJY-UHFFFAOYSA-N | |
methyl 2-(2,4-dichlorophenyl)acetate | |
2734106 | |
≥99.0% (GC) |
55954-23-9 | |
153°C | |
MFCD00018381 | |
methyl 2,4-dichlorophenylacetate, methyl 2-2,4-dichlorophenyl acetate, 2,4-dichlorophenylacetic acid methyl ester, benzeneacetic acid, 2,4-dichloro-, methyl ester, methyl2,4-dichlorophenylacetate, dcaa methyl ester, acmc-209lq7, methyl 2,4-dichlorobenzeneacetate, 2,4-dichlorophenylaceticacidmethylester, 2,4-dichlorobenzeneacetic acid methyl ester | |
COC(=O)CC1=C(C=C(C=C1)Cl)Cl | |
219.061 | |
219.06 | |
Liquid |
Chemical Identifiers
55954-23-9 | |
219.061 | |
SRCVNBFTEARRJY-UHFFFAOYSA-N | |
2734106 | |
COC(=O)CC1=C(C=C(C=C1)Cl)Cl |
C9H8Cl2O2 | |
MFCD00018381 | |
methyl 2,4-dichlorophenylacetate, methyl 2-2,4-dichlorophenyl acetate, 2,4-dichlorophenylacetic acid methyl ester, benzeneacetic acid, 2,4-dichloro-, methyl ester, methyl2,4-dichlorophenylacetate, dcaa methyl ester, acmc-209lq7, methyl 2,4-dichlorobenzeneacetate, 2,4-dichlorophenylaceticacidmethylester, 2,4-dichlorobenzeneacetic acid methyl ester | |
methyl 2-(2,4-dichlorophenyl)acetate |
Safety and Handling
TSCA : No