missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2,4-dichlorobenzoate, 97%

$53.11 - $149.45

Chemical Identifiers

CAS 35112-28-8
Molecular Formula C8H6Cl2O2
Formula Weight 205.04
MDL Number MFCD00085117
Synonym 2,4-Dichlorobenzoic acid methyl ester
Chemical Name or Material Methyl 2,4-dichlorobenzoate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2058309 SDS Alfa Aesar™
B2058309
10g Each for $149.45
AAB2058388 SDS Alfa Aesar™
B2058388
2.5g Each for $53.11
Specifications Description & Specifications

Chemical Identifiers

35112-28-8
205.04
2,4-Dichlorobenzoic acid methyl ester
C8H6Cl2O2
MFCD00085117
Methyl 2,4-dichlorobenzoate

Specifications

35112-28-8
205.04
2,4-Dichlorobenzoic acid methyl ester
10g
Methyl 2,4-dichlorobenzoate
C8H6Cl2O2
MFCD00085117
131° to 133°C (15mmHg)
110°C (230°F)
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning