missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2,4,6-trihydroxybenzoate, 98%

Manufacturer:  Alfa Aesar™ A1230514

 View more versions of this product

Catalog No. AAA1230514


 

Specifications

Specifications

Methyl 2,4,6-trihydroxybenzoate
98%
MFCD00013969
2,4,6-Trihydroxybenzoic acid methyl ester
COC(=O)C1=C(C=C(C=C1O)O)O
175°C to 179°C
76600
184.15
3147-39-5
C8H8O5
2837819
AQDIJIAUYXOCGX-UHFFFAOYSA-N
methyl 2,4,6-trihydroxybenzoate
184.147
25g

Chemical Identifiers

3147-39-5
184.147
AQDIJIAUYXOCGX-UHFFFAOYSA-N
76600
C8H8O5
MFCD00013969
2,4,6-Trihydroxybenzoic acid methyl ester
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents