missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2,3-dichlorophenylacetate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC371320250

 View more versions of this product

Catalog No. AC371320250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Methyl 2, 3-dichlorophenylacetate
25g
Authentic
1.5420 to 1.5440 (20°C, 589nm)
≥97.5%
C9H8Cl2O2
MFCD04112743
Clear colorless
10328-87-7
Glass bottle
Liquid
98%
219.07

Chemical Identifiers

10328-87-7
C9H8Cl2O2
MFCD04112743
≥97.5%
98%
219.07
Methyl 2, 3-dichlorophenylacetate

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.