Please login to your online account to display your discounted pricing

Methyl 2,3-dichlorophenylacetate, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC371320050

 View more versions of this product

Catalog No. AC371320050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 219.07
Quantity 5g
Color Clear colorless
Refractive Index 1.5420 to 1.5440 (20°C, 589nm)
Assay Percent Range 98%
CAS 10328-87-7
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Chemical Name or Material Methyl 2, 3-dichlorophenylacetate
Molecular Formula C9H8Cl2O2
Infrared Spectrum Authentic
MDL Number MFCD04112743

Specifications

Chemical Name or Material Methyl 2, 3-dichlorophenylacetate
CAS 10328-87-7
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H8Cl2O2
Formula Weight 219.07
MDL Number MFCD04112743
Accessories

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents