missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2,3-dichlorophenylacetate, 98%, ACROS Organics™

Chemical Identifiers

CAS 10328-87-7
Molecular Formula C9H8Cl2O2
Molecular Weight (g/mol) 219.07
MDL Number MFCD04112743
InChI Key XUMYFVFSYKMRDF-UHFFFAOYSA-N
Synonym acmc-1c9ir, benzeneacetic acid, 2,3-dichloro-, methyl ester, methyl 2,3-dichlorophenylacetate, methyl 2-2,3-dichlorophenyl acetate, methyl2,3-dichlorophenylacetate
PubChem CID 2734760
IUPAC Name methyl 2-(2,3-dichlorophenyl)acetate
SMILES COC(=O)CC1=C(C(=CC=C1)Cl)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC371320010
SDS View Documents
Thermo Fisher Scientific
AC371320010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC371320050
SDS View Documents
Thermo Fisher Scientific
AC371320050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC371320250
SDS View Documents
Thermo Fisher Scientific
AC371320250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

10328-87-7
219.07
XUMYFVFSYKMRDF-UHFFFAOYSA-N
2734760
COC(=O)CC1=C(C(=CC=C1)Cl)Cl
C9H8Cl2O2
MFCD04112743
acmc-1c9ir, benzeneacetic acid, 2,3-dichloro-, methyl ester, methyl 2,3-dichlorophenylacetate, methyl 2-2,3-dichlorophenyl acetate, methyl2,3-dichlorophenylacetate
methyl 2-(2,3-dichlorophenyl)acetate

Specifications

Authentic
1.5420 to 1.5440
1g
97.5% min. (GC)
MFCD04112743
XUMYFVFSYKMRDF-UHFFFAOYSA-N
methyl 2-(2,3-dichlorophenyl)acetate
2734760
Liquid
Methyl 2,3-dichlorophenylacetate
Glass bottle
Colorless
10328-87-7
C9H8Cl2O2
acmc-1c9ir, benzeneacetic acid, 2,3-dichloro-, methyl ester, methyl 2,3-dichlorophenylacetate, methyl 2-2,3-dichlorophenyl acetate, methyl2,3-dichlorophenylacetate
COC(=O)CC1=C(C(=CC=C1)Cl)Cl
219.07
219.07
98%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

SDS