missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2,3,6-Tri-O-benzoyl-alpha-D-galactopyranoside, TCI America™

$160.42 - $2,710.33

Chemical Identifiers

CAS 3601-36-3
Molecular Formula C28H26O9
Molecular Weight (g/mol) 506.507
MDL Number MFCD06200842
InChI Key WXFFEILSURAFKL-SUWSLWCISA-N
Synonym methyl 2,3,6-tri-o-benzoyl-alpha-d-galactopyranoside, 2s,3r,4s,5s,6r-6-benzoyloxy methyl-5-hydroxy-2-methoxytetrahydro-2h-pyran-3,4-diyl dibenzoate, methyl d-galactopyranoside 2,3,6-tribenzoate, methyl 2,3,6-tri-o-benzoyl-a-d-galactopyranoside, methyl 2,3,6-tri-o-benzoyl-, a-d-galactopyranoside, methyl 2-o,3-o,6-o-tribenzoyl-alpha-d-galactopyranoside, 2s,3r,4s,5s,6r-4-benzoyloxy-6-benzoyloxy methyl-5-hydroxy-2-methoxyoxan-3-yl benzoate, 2r,3s,4s,5r,6s-4,5-bis benzoyloxy-3-hydroxy-6-methoxyoxan-2-yl methyl benzoate
PubChem CID 10929259
IUPAC Name [(2R,3S,4S,5R,6S)-4,5-dibenzoyloxy-3-hydroxy-6-methoxyoxan-2-yl]methyl benzoate
SMILES COC1C(C(C(C(O1)COC(=O)C2=CC=CC=C2)O)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M19335G View Documents TCI America
M19335G
5g Each for $160.42
 
M1933250G View Documents TCI America
M1933250G
250g Each for $2,710.33
 
M193325G View Documents TCI America
M193325G
25g Each for $455.28
 
Specifications

Chemical Identifiers

3601-36-3
506.507
WXFFEILSURAFKL-SUWSLWCISA-N
10929259
COC1C(C(C(C(O1)COC(=O)C2=CC=CC=C2)O)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4
C28H26O9
MFCD06200842
methyl 2,3,6-tri-o-benzoyl-alpha-d-galactopyranoside, 2s,3r,4s,5s,6r-6-benzoyloxy methyl-5-hydroxy-2-methoxytetrahydro-2h-pyran-3,4-diyl dibenzoate, methyl d-galactopyranoside 2,3,6-tribenzoate, methyl 2,3,6-tri-o-benzoyl-a-d-galactopyranoside, methyl 2,3,6-tri-o-benzoyl-, a-d-galactopyranoside, methyl 2-o,3-o,6-o-tribenzoyl-alpha-d-galactopyranoside, 2s,3r,4s,5s,6r-4-benzoyloxy-6-benzoyloxy methyl-5-hydroxy-2-methoxyoxan-3-yl benzoate, 2r,3s,4s,5r,6s-4,5-bis benzoyloxy-3-hydroxy-6-methoxyoxan-2-yl methyl benzoate
[(2R,3S,4S,5R,6S)-4,5-dibenzoyloxy-3-hydroxy-6-methoxyoxan-2-yl]methyl benzoate

Specifications

White
5g
C28H26O9
methyl 2,3,6-tri-o-benzoyl-alpha-d-galactopyranoside, 2s,3r,4s,5s,6r-6-benzoyloxy methyl-5-hydroxy-2-methoxytetrahydro-2h-pyran-3,4-diyl dibenzoate, methyl d-galactopyranoside 2,3,6-tribenzoate, methyl 2,3,6-tri-o-benzoyl-a-d-galactopyranoside, methyl 2,3,6-tri-o-benzoyl-, a-d-galactopyranoside, methyl 2-o,3-o,6-o-tribenzoyl-alpha-d-galactopyranoside, 2s,3r,4s,5s,6r-4-benzoyloxy-6-benzoyloxy methyl-5-hydroxy-2-methoxyoxan-3-yl benzoate, 2r,3s,4s,5r,6s-4,5-bis benzoyloxy-3-hydroxy-6-methoxyoxan-2-yl methyl benzoate
COC1C(C(C(C(O1)COC(=O)C2=CC=CC=C2)O)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4
506.507
506.51
≥98.0% (HPLC)
143°C
3601-36-3
MFCD06200842
WXFFEILSURAFKL-SUWSLWCISA-N
[(2R,3S,4S,5R,6S)-4,5-dibenzoyloxy-3-hydroxy-6-methoxyoxan-2-yl]methyl benzoate
10929259
Crystal-Powder at 20°C
Methyl 2,3,6-Tri-O-benzoyl-alpha-D-galactopyranoside
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator