missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2,3,4,6-Tetra-O-acetyl-1-thio-beta-D-glucopyranoside, TCI America™

$255.14 - $864.73

Chemical Identifiers

CAS 13350-45-3
Molecular Formula C15H22O9S
Molecular Weight (g/mol) 378.392
InChI Key XWFUCHLBRWBKGN-QMIVOQANSA-N
PubChem CID 11164827
IUPAC Name [(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-methylsulfanyloxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)SC)OC(=O)C)OC(=O)C)OC(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M16821G View Documents TCI America
M16821G
1g Each for $255.14
 
M16825G View Documents TCI America
M16825G
5g Each for $864.73
 
Specifications

Chemical Identifiers

13350-45-3
378.392
11164827
CC(=O)OCC1C(C(C(C(O1)SC)OC(=O)C)OC(=O)C)OC(=O)C
C15H22O9S
XWFUCHLBRWBKGN-QMIVOQANSA-N
[(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-methylsulfanyloxan-2-yl]methyl acetate

Specifications

White
1g
C15H22O9S
CC(=O)OCC1C(C(C(C(O1)SC)OC(=O)C)OC(=O)C)OC(=O)C
378.392
378.39
≥98.0% (GC)
91°C
13350-45-3
XWFUCHLBRWBKGN-QMIVOQANSA-N
[(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-methylsulfanyloxan-2-yl]methyl acetate
11164827
Crystal-Powder at 20°C
Methyl 2,3,4,6-Tetra-O-acetyl-1-thio-beta-D-glucopyranoside
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator