missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2-(2,6-Dichloroanilino)phenylacetate, TCI America™

$125.28 - $375.84

Chemical Identifiers

CAS 15307-78-5
Molecular Formula C15H13Cl2NO2
Molecular Weight (g/mol) 310.174
MDL Number MFCD08273811
InChI Key VETACGBDFVVKGZ-UHFFFAOYSA-N
Synonym 2-(2,6-Dichloroanilino)phenylacetic Acid Methyl Ester, Diclofenac Methyl Ester
PubChem CID 519102
IUPAC Name methyl 2-[2-(2,6-dichloroanilino)phenyl]acetate
SMILES COC(=O)CC1=CC=CC=C1NC2=C(C=CC=C2Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M2903200MG View Documents TCI America
M2903200MG
200mg Each for $125.28
 
M29031G View Documents TCI America
M29031G
1g Each for $375.84
 
Specifications

Chemical Identifiers

15307-78-5
310.174
VETACGBDFVVKGZ-UHFFFAOYSA-N
519102
COC(=O)CC1=CC=CC=C1NC2=C(C=CC=C2Cl)Cl
C15H13Cl2NO2
MFCD08273811
2-(2,6-Dichloroanilino)phenylacetic Acid Methyl Ester, Diclofenac Methyl Ester
methyl 2-[2-(2,6-dichloroanilino)phenyl]acetate

Specifications

Undesignated
200mg
C15H13Cl2NO2
2-(2,6-Dichloroanilino)phenylacetic Acid Methyl Ester, Diclofenac Methyl Ester
COC(=O)CC1=CC=CC=C1NC2=C(C=CC=C2Cl)Cl
310.174
310.17
≥97.0% (GC,N)
102°C
15307-78-5
MFCD08273811
VETACGBDFVVKGZ-UHFFFAOYSA-N
methyl 2-[2-(2,6-dichloroanilino)phenyl]acetate
519102
Crystal-Powder at 20°C
Methyl 2-(2,6-Dichloroanilino)phenylacetate
Safety and Handling

Safety and Handling

TSCA : No