missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 2,2,3,3,3-Pentafluoropropyl Ether, TCI America™

$111.53 - $362.09

Chemical Identifiers

CAS 378-16-5
Molecular Formula C4H5F5O
Molecular Weight (g/mol) 164.075
MDL Number MFCD00236115
InChI Key ZYAMKYAPIQPWQR-UHFFFAOYSA-N
PubChem CID 2776015
IUPAC Name 1,1,1,2,2-pentafluoro-3-methoxypropane
SMILES COCC(C(F)(F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M25005G View Documents TCI America
M25005G
5g Each for $362.09
 
M25001G View Documents TCI America
M25001G
1g Each for $111.53
 
Specifications

Chemical Identifiers

378-16-5
164.075
ZYAMKYAPIQPWQR-UHFFFAOYSA-N
1,1,1,2,2-pentafluoro-3-methoxypropane
C4H5F5O
MFCD00236115
2776015
COCC(C(F)(F)F)(F)F

Specifications

48°C
5g
C4H5F5O
3271
COCC(C(F)(F)F)(F)F
164.075
164.08
≥98.0% (GC)
Colorless
378-16-5
MFCD00236115
ZYAMKYAPIQPWQR-UHFFFAOYSA-N
1,1,1,2,2-pentafluoro-3-methoxypropane
2776015
Clear Liquid at 20°C
Methyl 2,2,3,3,3-Pentafluoropropyl Ether
Safety and Handling

Safety and Handling

TSCA : No