Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 2-(2,2,2-trimethylacetamido)nicotinate

$405.38 - $405.38

Chemical Identifiers

Chemical Name or Material Methyl 2-(2,2,2-trimethylacetamido)nicotinate
CAS 773875-91-5
Molecular Formula C12H16N2O3
Formula Weight 236.27g/mol
MDL Number MFCD06204124
Synonym Methyl 2-(pivaloylamino)nicotinate; 2-(2,2,2-Trimethyacetamido)pyridine-3-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5001206

 
alfa aesar™
H50012-06
5g Each for $405.38
Description & Specifications

Specifications

Quantity 5g
Quantity 5g
Chemical Name or Material Methyl 2-(2,2,2-trimethylacetamido)nicotinate
CAS 773875-91-5
Molecular Formula C12H16N2O3
Formula Weight 236.27g/mol
MDL Number MFCD06204124
Synonym Methyl 2-(pivaloylamino)nicotinate; 2-(2,2,2-Trimethyacetamido)pyridine-3-carboxylic acid methyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.