Please login to your online account to display your discounted pricing

Methyl 1H-indole-2-carboxylate, 97%, Maybridge

$13.49 - $13.49

Chemical Identifiers

Chemical Name or Material Methyl 1H-indole-2-carboxylate
CAS 1202-04-6
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C10H9NO2
Formula Weight 175.19
MDL Number MFCD00460779
Synonym MFCD00460779
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO00996DA

  SDS
fisher scientific company
MO00996DA
Amber glass bottle 1g Each for $13.49

MO00996DE

 
fisher scientific company
MO00996DE
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 175.19
Quantity 1g
Assay 97%
CAS Min % 97.0
Packaging Amber glass bottle
Assay Percent Range 97%
Chemical Name or Material Methyl 1H-indole-2-carboxylate
MDL Number MFCD00460779
Synonym MFCD00460779
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Health Hazard 1 GHS Signal Word: Warning
CAS 1202-04-6
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C10H9NO2
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.