missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl-1H-benzotriazole (mixture), TCI America™

Manufacturer:  TCI America M069725G

 View more versions of this product

Catalog No. M069725G



Specifications

Specifications

Methyl-1H-benzotriazole (mixture)
87°C
29385-43-1
MFCD00167158
Crystal-Powder at 20°C
White-Yellow
25g
C7H7N3
133.15
≥98.0% (GC)

Chemical Identifiers

29385-43-1
C7H7N3
MFCD00167158
Safety and Handling

Safety and Handling

RTECSNumber : DM1300000

TSCA : No

Recommended Storage : Refrigerator

Documents