Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 1-trifluoromethyl-1-cyclopropanecarboxylate, 96%

Chemical Identifiers

Chemical Name or Material Methyl 1-trifluoromethyl-1-cyclopropanecarboxylate
CAS 208242-25-5
Molecular Formula C6H7F3O2
Formula Weight 168.12
MDL Number MFCD06801321
UN Number UN3272
Synonym 1-Trifluoromethyl-1-cyclopropanecarboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2575703

  SDS
alfa aesar™
H2575703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH2575706

  SDS
alfa aesar™
H2575706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH25757MD

  SDS
alfa aesar™
H25757MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 168.12
Chemical Name or Material Methyl 1-trifluoromethyl-1-cyclopropanecarboxylate
Assay 96%
Molecular Formula C6H7F3O2
MDL Number MFCD06801321
CAS 208242-25-5
UN Number UN3272
Synonym 1-Trifluoromethyl-1-cyclopropanecarboxylic acid methyl ester
Health Hazard 1 Warning
Boiling Point 115°C
Quantity 1g
Flash Point 42°C (108°F)
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.