missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 1-(tert-Butoxycarbonyl)-4-piperidinecarboxylate, TCI America™

$19.86 - $58.06

Chemical Identifiers

CAS 124443-68-1
Molecular Formula C12H21NO4
Molecular Weight (g/mol) 243.303
MDL Number MFCD02183584
InChI Key PTZNCCIULVXFIJ-UHFFFAOYSA-N
Synonym 1-Boc-4-piperidinecarboxylic Acid Methyl Ester, 1-(tert-Butoxycarbonyl)-4-piperidinecarboxylic Acid Methyl Ester, 1-tert-Butyl 4-Methyl Piperidine-1,4-dicarboxylate, Methyl 1-Boc-4-piperidinecarboxylate, Piperidine-1,4-dicarboxylic Acid 1-tert-Butyl 4-Met
PubChem CID 392870
IUPAC Name 1-O-tert-butyl 4-O-methyl piperidine-1,4-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M22275G View Documents TCI America
M22275G
5g Each for $58.06
 
M22271G View Documents TCI America
M22271G
1g Each for $19.86
 
Specifications

Chemical Identifiers

124443-68-1
243.303
PTZNCCIULVXFIJ-UHFFFAOYSA-N
392870
CC(C)(C)OC(=O)N1CCC(CC1)C(=O)OC
C12H21NO4
MFCD02183584
1-Boc-4-piperidinecarboxylic Acid Methyl Ester, 1-(tert-Butoxycarbonyl)-4-piperidinecarboxylic Acid Methyl Ester, 1-tert-Butyl 4-Methyl Piperidine-1,4-dicarboxylate, Methyl 1-Boc-4-piperidinecarboxylate, Piperidine-1,4-dicarboxylic Acid 1-tert-Butyl 4-Met
1-O-tert-butyl 4-O-methyl piperidine-1,4-dicarboxylate

Specifications

White-Yellow
5g
C12H21NO4
1-Boc-4-piperidinecarboxylic Acid Methyl Ester, 1-(tert-Butoxycarbonyl)-4-piperidinecarboxylic Acid Methyl Ester, 1-tert-Butyl 4-Methyl Piperidine-1,4-dicarboxylate, Methyl 1-Boc-4-piperidinecarboxylate, Piperidine-1,4-dicarboxylic Acid 1-tert-Butyl 4-Met
CC(C)(C)OC(=O)N1CCC(CC1)C(=O)OC
243.303
243.30
≥98.0% (GC)
35°C
124443-68-1
MFCD02183584
PTZNCCIULVXFIJ-UHFFFAOYSA-N
1-O-tert-butyl 4-O-methyl piperidine-1,4-dicarboxylate
392870
Crystal-Lump at 20°C
Methyl 1-(tert-Butoxycarbonyl)-4-piperidinecarboxylate
Safety and Handling

Safety and Handling

TSCA : No