missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylate, TCI America™

$36.67 - $137.50

Chemical Identifiers

CAS 148763-41-1
Molecular Formula C12H21NO4
Molecular Weight (g/mol) 243.303
MDL Number MFCD06795926
InChI Key LYYJQMCPEHXOEW-UHFFFAOYSA-N
Synonym Methyl 1-Boc-3-piperidinecarboxylate, 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylic Acid Methyl Ester, 1-Boc-3-piperidinecarboxylic Acid Methyl Ester, 1-tert-Butyl 3-Methyl Piperidine-1,3-dicarboxylate, Piperidine-1,3-dicarboxylic Acid 1-tert-Butyl 3-Met
PubChem CID 23590652
IUPAC Name 1-O-tert-butyl 3-O-methyl piperidine-1,3-dicarboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M291825G View Documents TCI America
M291825G
25g Each for $137.50
 
M29185G View Documents TCI America
M29185G
5g Each for $36.67
 
Specifications

Chemical Identifiers

148763-41-1
243.303
LYYJQMCPEHXOEW-UHFFFAOYSA-N
23590652
CC(C)(C)OC(=O)N1CCCC(C1)C(=O)OC
C12H21NO4
MFCD06795926
Methyl 1-Boc-3-piperidinecarboxylate, 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylic Acid Methyl Ester, 1-Boc-3-piperidinecarboxylic Acid Methyl Ester, 1-tert-Butyl 3-Methyl Piperidine-1,3-dicarboxylate, Piperidine-1,3-dicarboxylic Acid 1-tert-Butyl 3-Met
1-O-tert-butyl 3-O-methyl piperidine-1,3-dicarboxylate

Specifications

White-Yellow
25g
C12H21NO4
Methyl 1-Boc-3-piperidinecarboxylate, 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylic Acid Methyl Ester, 1-Boc-3-piperidinecarboxylic Acid Methyl Ester, 1-tert-Butyl 3-Methyl Piperidine-1,3-dicarboxylate, Piperidine-1,3-dicarboxylic Acid 1-tert-Butyl 3-Met
CC(C)(C)OC(=O)N1CCCC(C1)C(=O)OC
243.303
243.30
≥98.0% (GC)
49°C
148763-41-1
MFCD06795926
LYYJQMCPEHXOEW-UHFFFAOYSA-N
1-O-tert-butyl 3-O-methyl piperidine-1,3-dicarboxylate
23590652
Crystal-Powder at 20°C
Methyl 1-(tert-Butoxycarbonyl)-3-piperidinecarboxylate
Safety and Handling

Safety and Handling

TSCA : No