missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 1-naphthylacetate, 97%

$30.78 - $303.24

Chemical Identifiers

CAS 2876-78-0
Molecular Formula C13H12O2
Formula Weight 200.24
MDL Number MFCD00004045
Beilstein 1956293
Synonym Methyl naphthalene-1-acetate; 1-Naphthylacetic acid methyl ester
Chemical Name or Material Methyl 1-naphthylacetate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1312606 SDS Alfa Aesar™
A1312606
5g Each for $30.78
AAA1312614 SDS Alfa Aesar™
A1312614
25g Each for $93.43
AAA1312622 SDS Alfa Aesar™
A1312622
100g Each for $303.24
Specifications

Description

Applications
Methyl 1-naphthylacetate, is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Sparingly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Description & Specifications

Chemical Identifiers

2876-78-0
200.24
1956293
Methyl 1-naphthylacetate
C13H12O2
MFCD00004045
Methyl naphthalene-1-acetate; 1-Naphthylacetic acid methyl ester

Specifications

2876-78-0
200.24
1956293
155° to 157°C (9mmHg)
>110°C (230°F)
97%
C13H12O2
MFCD00004045
Methyl naphthalene-1-acetate; 1-Naphthylacetic acid methyl ester
5g
Methyl 1-naphthylacetate

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : QJ1225000