missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 1-Methylpyrrole-2-acetate 97.0+%, TCI America™

Manufacturer:  TCI America M084625ML

 View more versions of this product

Catalog No. M084625ML



Specifications

Specifications

Methyl 1-Methylpyrrole-2-acetate
Undesignated
51856-79-2
MFCD00009752
CGYVDYLHQYNGFC-UHFFFAOYSA-N
methyl 2-(1-methylpyrrol-2-yl)acetate
103992
Clear Liquid at 20°C
85°C
25mL
C8H11NO2
1-Methylpyrrole-2-acetic Acid Methyl Ester
CN1C=CC=C1CC(=O)OC
153.181
153.18
≥97.0% (GC)

Chemical Identifiers

51856-79-2
153.181
CGYVDYLHQYNGFC-UHFFFAOYSA-N
103992
CN1C=CC=C1CC(=O)OC
C8H11NO2
MFCD00009752
1-Methylpyrrole-2-acetic Acid Methyl Ester
methyl 2-(1-methylpyrrol-2-yl)acetate
Safety and Handling

Safety and Handling

TSCA : No

Documents