missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 1-Methylpyrrole-2-acetate, TCI America™

$105.42 - $311.67

Chemical Identifiers

CAS 51856-79-2
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.181
MDL Number MFCD00009752
InChI Key CGYVDYLHQYNGFC-UHFFFAOYSA-N
Synonym 1-Methylpyrrole-2-acetic Acid Methyl Ester
PubChem CID 103992
IUPAC Name methyl 2-(1-methylpyrrol-2-yl)acetate
SMILES CN1C=CC=C1CC(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M08465ML View Documents TCI America
M08465ML
5mL Each for $105.42
 
M084625ML View Documents TCI America
M084625ML
25mL Each for $311.67
 
Specifications

Chemical Identifiers

51856-79-2
153.181
CGYVDYLHQYNGFC-UHFFFAOYSA-N
103992
CN1C=CC=C1CC(=O)OC
C8H11NO2
MFCD00009752
1-Methylpyrrole-2-acetic Acid Methyl Ester
methyl 2-(1-methylpyrrol-2-yl)acetate

Specifications

85°C
5mL
C8H11NO2
1-Methylpyrrole-2-acetic Acid Methyl Ester
CN1C=CC=C1CC(=O)OC
153.181
153.18
≥97.0% (GC)
Undesignated
51856-79-2
MFCD00009752
CGYVDYLHQYNGFC-UHFFFAOYSA-N
methyl 2-(1-methylpyrrol-2-yl)acetate
103992
Clear Liquid at 20°C
Methyl 1-Methylpyrrole-2-acetate
Safety and Handling

Safety and Handling

TSCA : No