missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 1-methylimidazole-5-carboxylate, 98%

Manufacturer:  Alfa Aesar™ 4468803

 View more versions of this product

Catalog No. AA4468803



Specifications

Specifications

Methyl 1-methylimidazole-5-carboxylate
56°C to 58°C
17289-20-2
MFCD01567300
AKDPLDCXQNEMCL-UHFFFAOYSA-N
methyl 3-methylimidazole-4-carboxylate
2736896
Powder
Yellow
1g
C6H8N2O2
1-methyl-1h-imidazole-5-carboxylic acid methyl ester
CN1C=NC=C1C(=O)OC
140.142
140.14
98%

Chemical Identifiers

17289-20-2
140.142
AKDPLDCXQNEMCL-UHFFFAOYSA-N
2736896
CN1C=NC=C1C(=O)OC
C6H8N2O2
MFCD01567300
1-methyl-1h-imidazole-5-carboxylic acid methyl ester
methyl 3-methylimidazole-4-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents