missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 1-methylcyclopropyl ketone, 96%, Thermo Scientific Chemicals

 

Supplier:  Thermo Scientific Chemicals L0539406

Catalog No. AAL0539406



Description

Description

Methyl 1-methylcyclopropyl ketone is used as a pharmaceutical intermediate and an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 1-methylcyclopropyl ketone is used as a pharmaceutical intermediate and an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly
Specifications

Specifications

Methyl 1-methylcyclopropyl ketone
0.896
23°C (73°F)
1.434
5 g
2038600
Insoluble in water.
CC(=O)C1(CC1)C
98.145
98.15
1567-75-5
126°C to 128°C
C6H10O
MFCD00010245
UN1224
methyl 1-methylcyclopropyl ketone, 1-1-methylcyclopropyl ethan-1-one, 1-1-methylcyclopropyl ethanone, 1-acetyl-1-methylcyclopropane, ethanone, 1-1-methylcyclopropyl, 1-methylcyclopropyl methyl ketone, methyl 1-methyl cyclopropyl ketone, 1-1-methyl-cyclopropyl-ethanone, methyl1-methylcyclopropylketone
OQBCJXUAQQMTRW-UHFFFAOYSA-N
1-(1-methylcyclopropyl)ethanone
74067
96%

Chemical Identifiers

1567-75-5
98.145
OQBCJXUAQQMTRW-UHFFFAOYSA-N
74067
CC(=O)C1(CC1)C
C6H10O
MFCD00010245
methyl 1-methylcyclopropyl ketone, 1-1-methylcyclopropyl ethan-1-one, 1-1-methylcyclopropyl ethanone, 1-acetyl-1-methylcyclopropane, ethanone, 1-1-methylcyclopropyl, 1-methylcyclopropyl methyl ketone, methyl 1-methyl cyclopropyl ketone, 1-1-methyl-cyclopropyl-ethanone, methyl1-methylcyclopropylketone
1-(1-methylcyclopropyl)ethanone
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

EINECSNumber : 216-369-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only