Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 1-methylcyclopropyl ketone, 96%

Chemical Identifiers

Chemical Name or Material Methyl 1-methylcyclopropyl ketone
CAS 1567-75-5
Molecular Formula C6H10O
Formula Weight 98.15
MDL Number MFCD00010245
UN Number UN1224
Beilstein 2038600
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0539406

  SDS
alfa aesar™
L0539406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 98.15
Chemical Name or Material Methyl 1-methylcyclopropyl ketone
MDL Number MFCD00010245
Health Hazard 1 Warning
CAS 1567-75-5
Assay 96%
Molecular Formula C6H10O
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Beilstein 2038600
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
UN Number UN1224
Quantity 5g
Boiling Point 126° to 128°C
Flash Point 23°C (73°F)
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
Methyl 1-methylcyclopropyl ketone is used as a pharmaceutical intermediate and an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.