Please login to your online account to display your discounted pricing

Methyl 1-methylcyclopropyl ketone, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl 1-methylcyclopropyl ketone
Assay ≥94% (GC)
Assay Percent Range 95%
Molecular Formula C6H10O
Linear Formula CH3C3H4COCH3
Formula Weight 98.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC147630050

  SDS
thermo fisher scientific
147630050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC147630250

  SDS
thermo fisher scientific
147630250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC147631000

  SDS
thermo fisher scientific
AC147631000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94% (GC)
Physical Form Liquid
Formula Weight 98.14
Quantity 5g
Color Clear colorless to yellow
Density 0.895g/mL
Molecular Formula C6H10O
Refractive Index 1.4320 to 1.4350 (20°C, 589nm)
Infrared Spectrum Authentic
Linear Formula CH3C3H4COCH3
Assay Percent Range 95%
Packaging Glass bottle
Chemical Name or Material Methyl 1-methylcyclopropyl ketone
CAS 1567-75-5
Health Hazard 1 Warning
Safety and Handling

Warning