missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 1-methyl-5-oxopyrrolidine-3-carboxylate, 97%

Manufacturer:  Alfa Aesar™ H6357706

 View more versions of this product

Catalog No. AAH6357706



Specifications

Specifications

Methyl 1-methyl-5-oxopyrrolidine-3-carboxylate
59857-86-2
MFCD00015898
JFZRRXGXEHUJCI-UHFFFAOYSA-N
methyl 1-methyl-5-oxopyrrolidine-3-carboxylate
108402
Solid
5g
C7H11NO3
methyl 1-methyl-2-oxopyrrolidine-4-carboxylate, 1-methyl-5-oxo-3-pyrrolidinecarboxylic acid methyl ester, 3-pyrrolidinecarboxylic acid, 1-methyl-5-oxo-, methyl ester, 1-methyl-4-methyloxycarbonyl-2-pyrrolidone, 1-methyl-5-oxo-pyrrolidine-3-carboxylic acid methyl ester, 5-22-06-00040 beilstein handbook reference, 4-carbomethoxy-1-methyl-pyrrolidone, methyl n-methyl-2-pyrrolidone-4-carboxylate, methyl1-methyl-2-oxopyrrolidine-4-carboxylate, methyl 1-methyl-5-oxo-3-pyrrolidinecarboxylate
CN1CC(CC1=O)C(=O)OC
157.169
157.17
97%

Chemical Identifiers

59857-86-2
157.169
JFZRRXGXEHUJCI-UHFFFAOYSA-N
108402
CN1CC(CC1=O)C(=O)OC
C7H11NO3
MFCD00015898
methyl 1-methyl-2-oxopyrrolidine-4-carboxylate, 1-methyl-5-oxo-3-pyrrolidinecarboxylic acid methyl ester, 3-pyrrolidinecarboxylic acid, 1-methyl-5-oxo-, methyl ester, 1-methyl-4-methyloxycarbonyl-2-pyrrolidone, 1-methyl-5-oxo-pyrrolidine-3-carboxylic acid methyl ester, 5-22-06-00040 beilstein handbook reference, 4-carbomethoxy-1-methyl-pyrrolidone, methyl n-methyl-2-pyrrolidone-4-carboxylate, methyl1-methyl-2-oxopyrrolidine-4-carboxylate, methyl 1-methyl-5-oxo-3-pyrrolidinecarboxylate
methyl 1-methyl-5-oxopyrrolidine-3-carboxylate
Safety and Handling

Safety and Handling

RTECSNumber : UY0557000

Documents