Please login to your online account to display your discounted pricing

Methyl 1-hydroxy-2-naphthoate, Acros Organics


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC428470010

 View more versions of this product

Catalog No. AC428470010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Methyl 1-hydroxy-2-naphthoate
CAS 948-03-8
Packaging Glass bottle
Molecular Formula C12H10O3
Formula Weight 202.21
Quantity 1g

Specifications

Chemical Name or Material Methyl 1-hydroxy-2-naphthoate
CAS 948-03-8
Molecular Formula C12H10O3
Formula Weight 202.21