Please login to your online account to display your discounted pricing

Methyl 1-hydroxy-2-naphthoate, Acros Organics

Chemical Identifiers

Chemical Name or Material Methyl 1-hydroxy-2-naphthoate
CAS 948-03-8
Molecular Formula C12H10O3
Formula Weight 202.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC428470010

  SDS
thermo fisher scientific
AC428470010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC428470050

  SDS
thermo fisher scientific
AC428470050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Methyl 1-hydroxy-2-naphthoate
CAS 948-03-8
Packaging Glass bottle
Molecular Formula C12H10O3
Formula Weight 202.21
Quantity 1g