missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 1-cyclohexene-1-carboxylate, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC213710250

 View more versions of this product

Catalog No. AC213710250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Methyl 1-cyclohexene-1-carboxylate
Authentic
1.4755 to 1.4775 (20°C, 589nm)
25g
98%
C6H9CO2CH3
09, IV, 112
KXPWRCPEMHIZGU-UHFFFAOYSA-N
methyl cyclohexene-1-carboxylate
87647
Liquid
1.03g/mL
Glass bottle
Colorless
18448-47-0
C8H12O2
MFCD00001544
1-carbomethoxy cyclohexene, 1-cyclohexene-1-carboxylic acid, methyl ester, cyclohexene-1-carboxylic acid methyl ester, cyclohexenecarboxylic acid, methyl ester, methyl 1-cyclohexene-1-carboxylate, methyl 1-cyclohexenecarboxylate, methyl cyclohex-1-ene-1-carboxylate, methyl cyclohex-1-enecarboxylate, methyl1-cyclohexene-1-carboxylate, pubchem11031
COC(=O)C1=CCCCC1
140.182
140.18
≥97.5%

Chemical Identifiers

18448-47-0
140.182
KXPWRCPEMHIZGU-UHFFFAOYSA-N
87647
COC(=O)C1=CCCCC1
C8H12O2
MFCD00001544
1-carbomethoxy cyclohexene, 1-cyclohexene-1-carboxylic acid, methyl ester, cyclohexene-1-carboxylic acid methyl ester, cyclohexenecarboxylic acid, methyl ester, methyl 1-cyclohexene-1-carboxylate, methyl 1-cyclohexenecarboxylate, methyl cyclohex-1-ene-1-carboxylate, methyl cyclohex-1-enecarboxylate, methyl1-cyclohexene-1-carboxylate, pubchem11031
methyl cyclohexene-1-carboxylate
SDS Documents