missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Methyl 1-cyclohexene-1-carboxylate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 213710050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Methyl 1-cyclohexene-1-carboxylate | |
1.0300g/mL | |
73°C | |
97.5% min. (GC) | |
C8H12O2 | |
C6H9CO2CH3 | |
5g | |
1.03 | |
Solubility in water: insoluble. Other solubilities: soluble in organic solvents | |
COC(=O)C1=CCCCC1 | |
140.18 | |
140.18 |
18448-47-0 | |
190°C to 192°C | |
Authentic | |
Glass bottle | |
1.4755 to 1.4775 | |
MFCD00001544 | |
09, IV, 112 | |
methyl 1-cyclohexene-1-carboxylate, methyl cyclohex-1-enecarboxylate, methyl 1-cyclohexenecarboxylate, methyl cyclohex-1-ene-1-carboxylate, 1-cyclohexene-1-carboxylic acid, methyl ester, cyclohexenecarboxylic acid, methyl ester, cyclohexene-1-carboxylic acid methyl ester, methyl1-cyclohexene-1-carboxylate, pubchem11031, 1-carbomethoxy cyclohexene | |
KXPWRCPEMHIZGU-UHFFFAOYSA-N | |
methyl cyclohexene-1-carboxylate | |
87647 | |
98% |
Chemical Identifiers
18448-47-0 | |
140.18 | |
KXPWRCPEMHIZGU-UHFFFAOYSA-N | |
87647 | |
COC(=O)C1=CCCCC1 |
C8H12O2 | |
MFCD00001544 | |
methyl 1-cyclohexene-1-carboxylate, methyl cyclohex-1-enecarboxylate, methyl 1-cyclohexenecarboxylate, methyl cyclohex-1-ene-1-carboxylate, 1-cyclohexene-1-carboxylic acid, methyl ester, cyclohexenecarboxylic acid, methyl ester, cyclohexene-1-carboxylic acid methyl ester, methyl1-cyclohexene-1-carboxylate, pubchem11031, 1-carbomethoxy cyclohexene | |
methyl cyclohexene-1-carboxylate |
Safety and Handling
EINECSNumber : 242-331-5
RUO â Research Use Only