Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 1-chloronaphtho[2,1-b]thiophene-2-carboxylate, 96%

Chemical Identifiers

Chemical Name or Material Methyl 1-chloronaphtho[2,1-b]thiophene-2-carboxylate
CAS 85992-30-9
Formula Weight 276.74
MDL Number MFCD23380841
Synonym 3-Chloro-2-carboxynaphtho[2,1-b]thiophene methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3640103

 
alfa aesar™
H3640103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH36401MD

 
alfa aesar™
H36401MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 276.74
Chemical Name or Material Methyl 1-chloronaphtho[2,1-b]thiophene-2-carboxylate
Physical Form Powder
Assay 96%
MDL Number MFCD23380841
CAS 85992-30-9
Synonym 3-Chloro-2-carboxynaphtho[2,1-b]thiophene methyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Brown
Melting Point ≈135°C (dec.)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.