missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl (+/-)-1-benzyloxycarbonyl-4-Boc-piperazine-2-carboxylate

$92.66 - $706.53

Chemical Identifiers

CAS 126937-42-6
Molecular Formula C19H26N2O6
Formula Weight 378.43
MDL Number MFCD03426296
Synonym (+/-)-1-Benzyloxycarbonyl-4-Boc-piperazine-2-carboxylic acid methyl ester; Piperazine-1,2,4-tricarboxylic acid 1-benzyl 4-tert-butyl 2-methyl triester
Chemical Name or Material Methyl (+/-)-1-benzyloxycarbonyl-4-Boc-piperazine-2-carboxylate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2783203 SDS Alfa Aesar™
H2783203
1g Each for $232.43
AAH2783206 SDS Alfa Aesar™
H2783206
5g Each for $706.53
AAH27832MD SDS Alfa Aesar™
H27832MD
250mg Each for $92.66
Specifications Description & Specifications

Chemical Identifiers

126937-42-6
378.43
(+/-)-1-Benzyloxycarbonyl-4-Boc-piperazine-2-carboxylic acid methyl ester; Piperazine-1,2,4-tricarboxylic acid 1-benzyl 4-tert-butyl 2-methyl triester
C19H26N2O6
MFCD03426296
Methyl (+/-)-1-benzyloxycarbonyl-4-Boc-piperazine-2-carboxylate

Specifications

126937-42-6
378.43
(+/-)-1-Benzyloxycarbonyl-4-Boc-piperazine-2-carboxylic acid methyl ester; Piperazine-1,2,4-tricarboxylic acid 1-benzyl 4-tert-butyl 2-methyl triester
Methyl (+/-)-1-benzyloxycarbonyl-4-Boc-piperazine-2-carboxylate
C19H26N2O6
MFCD03426296
1g

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning