missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, 95%, Thermo Scientific Chemicals

$187.49 - $514.78

Chemical Identifiers

CAS 3939-01-3
MDL Number MFCD00012799
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2693014
View Documents Promotion Details
Thermo Scientific Chemicals
H2693014
25 g
Each for $187.49
 
AAH2693022
View Documents Promotion Details
Thermo Scientific Chemicals
H2693022
100 g
Each for $514.78
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3939-01-3
MFCD00012799

Specifications

3939-01-3
C14H18ClNO3
25 g
methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, methyl 1-benzyl-4-oxo-3-piperidinecarboxylate hydrochloride, methyl 1-benzyl-4-oxo-3-piperidine-carboxylate hydrochloride, 1-benzyl-3-carbomethoxy-4-piperidone hydrochloride, 1-benzyl-3-methoxycarbonyl-4-piperidone hydrochloride, methyl 1-benzyl-4-oxopiperidine-3-carboxylate hcl, 1-benzyl-3-methoxycarbonyl-4-piperidone hcl, pubchem12956, acmc-1cll5, ksc790s5l
COC(=O)C1CN(CCC1=O)CC2=CC=CC=C2.Cl
283.75
283.75
Methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride
∽185°C (decomposition)
MFCD00012799
Hygroscopic
BRADBAOVPACOQQ-UHFFFAOYNA-N
methyl 1-benzyl-4-oxopiperidine-3-carboxylate;hydrochloride
107479
95%
Safety and Handling

Safety and Handling

EINECSNumber : 223-522-2

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only