missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 1-Benzyl-4-oxo-3-piperidinecarboxylate Hydrochloride, TCI America™

$44.31 - $154.31

Chemical Identifiers

CAS 3939-01-3
Molecular Formula C14H18ClNO3
Molecular Weight (g/mol) 283.75
MDL Number MFCD00012799
InChI Key BRADBAOVPACOQQ-UHFFFAOYNA-N
Synonym methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, methyl 1-benzyl-4-oxo-3-piperidinecarboxylate hydrochloride, methyl 1-benzyl-4-oxo-3-piperidine-carboxylate hydrochloride, 1-benzyl-3-carbomethoxy-4-piperidone hydrochloride, 1-benzyl-3-methoxycarbonyl-4-piperidone hydrochloride, methyl 1-benzyl-4-oxopiperidine-3-carboxylate hcl, 1-benzyl-3-methoxycarbonyl-4-piperidone hcl, pubchem12956, acmc-1cll5, ksc790s5l
PubChem CID 107479
IUPAC Name methyl 1-benzyl-4-oxopiperidine-3-carboxylate;hydrochloride
SMILES COC(=O)C1CN(CCC1=O)CC2=CC=CC=C2.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M25315G View Documents TCI America
M25315G
5g Each for $44.31
 
M253125G View Documents TCI America
M253125G
25g Each for $154.31
 
Specifications

Chemical Identifiers

3939-01-3
283.75
BRADBAOVPACOQQ-UHFFFAOYNA-N
107479
COC(=O)C1CN(CCC1=O)CC2=CC=CC=C2.Cl
C14H18ClNO3
MFCD00012799
methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, methyl 1-benzyl-4-oxo-3-piperidinecarboxylate hydrochloride, methyl 1-benzyl-4-oxo-3-piperidine-carboxylate hydrochloride, 1-benzyl-3-carbomethoxy-4-piperidone hydrochloride, 1-benzyl-3-methoxycarbonyl-4-piperidone hydrochloride, methyl 1-benzyl-4-oxopiperidine-3-carboxylate hcl, 1-benzyl-3-methoxycarbonyl-4-piperidone hcl, pubchem12956, acmc-1cll5, ksc790s5l
methyl 1-benzyl-4-oxopiperidine-3-carboxylate;hydrochloride

Specifications

Undesignated
5g
C14H18ClNO3
methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, methyl 1-benzyl-4-oxo-3-piperidinecarboxylate hydrochloride, methyl 1-benzyl-4-oxo-3-piperidine-carboxylate hydrochloride, 1-benzyl-3-carbomethoxy-4-piperidone hydrochloride, 1-benzyl-3-methoxycarbonyl-4-piperidone hydrochloride, methyl 1-benzyl-4-oxopiperidine-3-carboxylate hcl, 1-benzyl-3-methoxycarbonyl-4-piperidone hcl, pubchem12956, acmc-1cll5, ksc790s5l
COC(=O)C1CN(CCC1=O)CC2=CC=CC=C2.Cl
283.75
283.75
≥98.0% (HPLC)
182°C
3939-01-3
MFCD00012799
BRADBAOVPACOQQ-UHFFFAOYNA-N
methyl 1-benzyl-4-oxopiperidine-3-carboxylate;hydrochloride
107479
Crystal-Powder at 20°C
Methyl 1-Benzyl-4-oxo-3-piperidinecarboxylate Hydrochloride
Safety and Handling

Safety and Handling

TSCA : No