missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Methyl 1-Benzyl-4-oxo-3-piperidinecarboxylate Hydrochloride 98.0+%, TCI America™

Manufacturer:  Tci America M253125G

 View more versions of this product

Catalog No. M253125G



Specifications

Specifications

Methyl 1-Benzyl-4-oxo-3-piperidinecarboxylate Hydrochloride
182°C
3939-01-3
MFCD00012799
BRADBAOVPACOQQ-UHFFFAOYNA-N
methyl 1-benzyl-4-oxopiperidine-3-carboxylate;hydrochloride
107479
Crystal-Powder at 20°C
Undesignated
25g
C14H18ClNO3
1-benzyl-3-carbomethoxy-4-piperidone hydrochloride, 1-benzyl-3-methoxycarbonyl-4-piperidone hcl, 1-benzyl-3-methoxycarbonyl-4-piperidone hydrochloride, acmc-1cll5, ksc790s5l, methyl 1-benzyl-4-oxo-3-piperidine-carboxylate hydrochloride, methyl 1-benzyl-4-oxo-3-piperidinecarboxylate hydrochloride, methyl 1-benzyl-4-oxopiperidine-3-carboxylate hcl, methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, pubchem12956
COC(=O)C1CN(CCC1=O)CC2=CC=CC=C2.Cl
283.75
283.75
≥98.0% (HPLC)

Chemical Identifiers

3939-01-3
283.75
BRADBAOVPACOQQ-UHFFFAOYNA-N
107479
COC(=O)C1CN(CCC1=O)CC2=CC=CC=C2.Cl
C14H18ClNO3
MFCD00012799
1-benzyl-3-carbomethoxy-4-piperidone hydrochloride, 1-benzyl-3-methoxycarbonyl-4-piperidone hcl, 1-benzyl-3-methoxycarbonyl-4-piperidone hydrochloride, acmc-1cll5, ksc790s5l, methyl 1-benzyl-4-oxo-3-piperidine-carboxylate hydrochloride, methyl 1-benzyl-4-oxo-3-piperidinecarboxylate hydrochloride, methyl 1-benzyl-4-oxopiperidine-3-carboxylate hcl, methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, pubchem12956
methyl 1-benzyl-4-oxopiperidine-3-carboxylate;hydrochloride
Safety and Handling

Safety and Handling

TSCA : No

Documents
Promotions

Promotions