Please login to your online account to display your discounted pricing

Methyl 1-benzyl-4-oxo-3-piperidine-carboxylate hydrochloride, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Methyl 1-benzyl-4-oxo-3-piperidine-carboxylate hydrochloride
Name Note 97%
CAS 744733
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C14H17NO3·HCl
Formula Weight 283.76
MDL Number MFCD00012799
Synonym 1-Benzyl-3-carbomethoxy-4-piperidone hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC210561000

  SDS
fisher scientific acros
210561000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC210560250

  SDS
fisher scientific acros
210560250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 283.76
Quantity 100g
Color White to pink
EINECS Number 223-522-2
CAS 744733
Chemical Name or Material Methyl 1-benzyl-4-oxo-3-piperidine-carboxylate hydrochloride
Infrared Spectrum Authentic
Name Note 97%
Packaging Glass bottle
Assay Percent Range 97%
Molecular Formula C14H17NO3·HCl
MDL Number MFCD00012799
Synonym 1-Benzyl-3-carbomethoxy-4-piperidone hydrochloride
Health Hazard 1 HYGROSCOPIC
Safety and Handling

HYGROSCOPIC