Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 1-aminocyclobutanecarboxylate hydrochloride, 95%

Chemical Identifiers

Chemical Name or Material Methyl 1-aminocyclobutanecarboxylate hydrochloride
CAS 215597-35-6
Formula Weight 165.62
MDL Number MFCD04115288
Beilstein 5615121
Synonym 1-Aminocyclobutanecarboxylic acid methyl ester hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6470103

  SDS
alfa aesar™
H6470103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6470106

  SDS
alfa aesar™
H6470106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH64701MD

  SDS
alfa aesar™
H64701MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 165.62
Synonym 1-Aminocyclobutanecarboxylic acid methyl ester hydrochloride
CAS 215597-35-6
MDL Number MFCD04115288
Physical Form Solid
Sensitivity Air Sensitive
Chemical Name or Material Methyl 1-aminocyclobutanecarboxylate hydrochloride
Beilstein 5615121
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Health Hazard 1 Warning
Assay 95%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

Applications
Used as a primary and secondary intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.