Please login to your online account to display your discounted pricing

Alfa Aesar™ Methyl 1-(6-chloro-3-pyridazinyl)piperidine-4-carboxylate

$131.07 - $131.07

Chemical Identifiers

Chemical Name or Material Methyl 1-(6-chloro-3-pyridazinyl)piperidine-4-carboxylate
CAS 339276-34-5
Molecular Formula C11H14ClN3O2
Formula Weight 255.71g/mol
MDL Number MFCD00664522
Synonym 1-(6-Chloro-3-pyridazinyl)piperidine-4-carboxylic acid methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5052303

 
alfa aesar™
H50523-03
Each for $131.07
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material Methyl 1-(6-chloro-3-pyridazinyl)piperidine-4-carboxylate
CAS 339276-34-5
Molecular Formula C11H14ClN3O2
Formula Weight 255.71g/mol
MDL Number MFCD00664522
Synonym 1-(6-Chloro-3-pyridazinyl)piperidine-4-carboxylic acid methyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.